Avantor® ACE®, HPLC ¹/₃₂" Column Connectors, Finger-Tight
Catalog # ACE-MC3210
Supplier: Avantor
Specifications
- Omschrijving:Microbore finger-tight HPLC column connector, 1/32", 6-40 thread
- Environmentally Preferable:Lage impact op de fabricage
- Milieu certificering:ISO 14001 : 2015 (530840 UM15)
- VPE:10
Specifications
About this item
PEEK ¹/₃₂" microbore HPLC column inlet / outlet connectors. These column connectors are recommended for ¹/₃₂" microbore HPLC column connections. They are compatible with both capillary (360 µm OD) and ¹/₃₂" connection tubing.
- Pressure rated to 400 bar (6000 psi)
- Reusable
- Compatible with all microbore columns with ¹/₃₂" and 6-40 threads