Orders LinkContactUsLinkComponent
Mitosis model
Mitosis model
Mitosis model
Catalogus # BINH1013868
Marketsource
Leverancier:   3B Scientific
Mitosis model
Catalogus # BINH1013868
Leverancier:  3B Scientific
Marketsource
Mitosis model
Mitosis model
Mitosis model
Price
405,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
This model series shows the following nine phases of mitosis on the basis of a typical mammal cell at an enlargement of 10000 times. The three-dimensional relief models are painted according to the usual colouring methods of microscopy, making the process of cell division easy to understand. The cell organelles are shown as if opened up in the lower part of the models.
  • 1. Interphase
  • 2. Prophase
  • 3. Early prometaphase
  • 4. Later prometaphase
  • 5. Metaphase
  • 6. Early anaphase
  • 7. Later anaphase
  • 8. Telophase
  • 9. Cytokinesis
Equipped with magnets at the back for teaching purposes, they can be easily arranged on a magnetic board in the classroom.
Bestelinformatie: Includes detailed description and handouts for your lessons.
Levering: Supplied in a storage system (400×600 mm), which can be fastened to the wall.
Specificaties
  • Omschrijving:
    Mitosis model
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.