Drigalski spreader
Drigalski spreader, Ø 5 mm, 50 mm blade width, transparant, 145 mm
Catalogus # MARI1800024
Leverancier: MARIENFELD
Ga naar:
Product Details & Documents
Glass.
- For bacteriological applications
- Facilitates uniform distribution and plating of bacteria material on solid nutrient media
Specificaties
- Kleur:transparant
- Omschrijving:Drigalski spreader, Ø 5 mm, 50 mm blade width
- Lengte:145 mm