Ga naar Rubriek
Ga naar Hoofdinhoud
Ga naar Voettekst
Geavanceerd
Geavanceerd
SDS
Certificaten
Gegevens van de fabrikant
0
0,00 €
Order Entry
Inloggen
/
Registreren
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Belgium
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
Microplaten
PCR-platen
PCR plates, 96-well, with full skirt, low profile well design, Multiply®
PCR plates, 96-well, with full skirt, low profile well design, Multiply®
PCR plates, 96-well, with full skirt, low profile well design, Multiply®
Catalog # SARS72.1980.202
Supplier:
SARSTEDT
Expand image
PCR plates, 96-well, with full skirt, low profile well design, Multiply®
Catalog # SARS72.1980.202
Supplier:
SARSTEDT
Specifications
Aant. wells:
96
Profiel:
Laag
Standring:
Volledige rand
Wellkleur:
helder
VPE:
200
Specifications
About this item
Product Details
Product Details