Hydrometer set
Complete set of 14 instruments, 1
Catalogus # AMARH809630
Leverancier: Amarell
Ga naar:
Product Details & Documents
Hydrometers with scale paper, colour code and type designation, weighted body, with (and without) thermometer inside the hydrometer body, round capillary, mercury filling suitable for government verification.
Delivery information: Supplied in a transparent plastic case.
Specificaties
- Omschrijving:Complete set of 14 instruments
- Indeling:1