Orders LinkContactUsLinkComponent
Rectangular spacers for SpectrumGX
Supplier:   PerkinElmer
Rectangular spacers for SpectrumGX
Supplier:  PerkinElmer
Certificates
Rectangular spacers for SpectrumGX
7 Options Available Below
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Product Family Options
Product Information
  • Omschrijving
    Voor
    Availability
    Price
  • Spacers, lead, rectangular, 0,025 mm thickness for semi-demountable cell
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST
  • Spacers, PTFE, rectangular, 0,05 mm thickness for semi-demountable cell
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 228,00 €
    228,00 €
    1 * 6 ST
       228,00 €
  • Spacers, PTFE, rectangular, 0,1 mm thickness for semi-demountable cell
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 758,00 €
    758,00 €
    1 * 6 ST
       758,00 €
  • Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 228,00 €
    228,00 €
    1 * 3 ST
       228,00 €
  • Spacers, PTFE, rectangular, 0,5 mm thickness for semi-demountable cell
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 228,00 €
    228,00 €
    1 * 3 ST
       228,00 €
  • Spacers, PTFE, rectangular, 1,0 mm thickness for semi-demountable cell
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 190,00 €
    190,00 €
    1 * 3 ST
       190,00 €
  • Assorted spacers for semi-demountable cell, includes two each of thickness 0,025, 0,05, 0,1, 0,2, 0,5, 1,0 mm spacers
    SpectrumGX
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 283,00 €
    283,00 €
    1 * 12 ST
       283,00 €