Orders LinkContactUsLinkComponent
DNA gel loading buffer 6X
6X DNA loading buffer, 5× 1 ml, 5
Catalogus # LONZ50655
Leverancier:   LONZA
DNA gel loading buffer 6X
Catalogus # LONZ50655
Leverancier:  LONZA
CAS-nummer:  
DNA gel loading buffer 6X
DNA gel loading buffer 6X
6X DNA loading buffer, 5× 1 ml, 5
CAS-nummer:  
Price
79,20 €
Log in voor beschikbaarheid
1 * 5 ST
Product Details & Documents
This buffer is a Ficoll based loading buffer containing Ficoll®, EDTA, xylene cyanol, and bromophenol blue. Ideal for separation of DNA fragments with improved resolution.
Specificaties
Specificaties
  • VPE:
    5
  • Omschrijving:
    6X DNA loading buffer, 5× 1 ml
Specificaties
Accessories
Productinformatie
  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • 6X DNA loading buffer, 5× 1 ml
    6X DNA loading buffer, 5× 1 ml
    LONZ50655
    5
    Log in voor beschikbaarheid
    1 * 5 ST - 79,20 €