Urine collection cup with screw lid
720-1333
- Omschrijving:Urine collection cup with screw lid + temp strip
- VPE:1
Urine collection cup with screw lid and temperature strip to show validity of sample. Suitable for use with all types of urine drugs of abuse test, apart from the integrated cup.
- Designed to accommodate all versions of multipanel test