Hair analysis set
Catalog # SUREHA
Supplier: SURESCREEN
Specifications
- Omschrijving:Hair analysis for drugs of abuse
- VPE:1
Specifications
About this item
Hair is a medium designed to analyse drug abuse over a period of three months. As the hair grows it records all the drugs that are in the body. Hair is easy to collect and cannot be altered.
- Standard hair test checks for the presence
- Amphetamines
- Cannabis (THC)
- Cocaine
- Methamphetamines
- Opiates
- PCP
Standard hair test checks for the presence of: Amphetamines; Cannabis (THC); Cocaine; Methamphetamines; Opiates; PCP.