Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 405)
ROCK609-446-002-05
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:DyLight® 405
- Host:Rabbit
- ImmunoFluorescentie:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Maat:500 µg
- VPE:500 µG
Type: Secondary
Antigen: IgG(Human)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight405
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human