Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Catalogus # ROCK704-406-002
Leverancier: Rockland Immunochemicals
Specificaties
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:Biotine
- ELISA:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- Isotype:
- Reactivity:Dog
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:No
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Antilichaamfragment:F(ab')2
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Opslagtemperatuur:–20 °C
- Shipping temperature:Ambient
- Immunogeen:Dog IgG whole molecule
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Maat:500 µg
- VPE:500 µG
Specificaties
Over dit artikel
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:1000
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Dog