Anti-MBP Chicken Polyclonal Antibody
Catalogus # ROCK200-901-D81
Leverancier: Rockland Immunochemicals
Specificaties
- Type antilichaam:Primair
- antigeen symbool:MBP
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- Host:Chicken
- ImmunoFluorescentie:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:No
- Formaat:Antibody fraction
- Formulier:Liquid
- Gen-ID:24547
- Antigeen synoniemen:Mbps
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Opslagtemperatuur:–20 °C
- Shipping temperature:Dry Ice
- Immunogeen:Anti-Myelin basic Protein Antibody was produced in chicken by repeated immunizations with three peptide sequences conserved in higher vertebrate MBP Proteins.
- Maat:100 µl
- VPE:100 µl
Specificaties
Over dit artikel
Anti-Myelin Basic Protein antibody is suitable for use in Western Blotting and IF. Expect a bands of approximately 18 kDa and 22 kDa in size corresponding to MBP Proteins in Western blots of Rat brain homogenate.
Recommended Dilutions: Immunofluorescence: 1:500; Western Blot: 1:5000
Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat