Anti-TP53BP2 Rabbit Polyclonal Antibody
100 µg
Catalogus # ROCK200-401-A19
Leverancier: Rockland Immunochemicals
Anti-TP53BP2 Rabbit Polyclonal Antibody
Catalogus # ROCK200-401-A19
Leverancier: Rockland Immunochemicals
Ga naar:
Product Details & Documents
This affinity purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 125 kDa in size corresponding to ASPP2 by western blotting.
Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: TP53BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
TDS Anti-TP53BP2 Rabbit Polyclonal Antibody
Technische fiche
Specificaties
- Maat:100 µg
- Western blot:Yes
- Clonality:Polyklonaal
- antigeen symbool:TP53BP2
- Purification:Protein A purified
- Type antilichaam:Primair
- Formaat:Antibody fraction
- Isotype:IgG
- Host:Rabbit
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Shipping temperature:Dry Ice
- ImmunoChemie:Yes
- Epitoop:Internal
- Cat. No.:ROCK200-401-A19
- Antigeen synoniemen:P53BP2|ASPP2|PPP1R13A|53BP2|BBP
- Gen-ID:7159
- Conjugatie:Onconjugeerd
- Reactivity:Human
- Opslagtemperatuur:–20 °C
- Kruisadsorptie:No
- Formulier:Liquid (sterile filtered)
- Immunogeen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of human ASPP2.
- ELISA:Yes



