Anti-IgG F(c) Goat Polyclonal Antibody
ROCK610-1122
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG F(c)
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Goat
- ImmunoChemie:Yes
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:No
- Formaat:Antibody fraction
- Formulier:Liquid (sterile filtered)
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Opslagtemperatuur:–20 °C
- Shipping temperature:Wet Ice
- Immunogeen:Mouse IgG F(c) fragment
- Purification:purified by immunoaffinity chromatography
- Maat:1 mg
- VPE:1 mg
Anti-Mouse IgG F(c) antibody is suitable for ELISA, Western Blot and immunochemistry applications. Antibody should be optimized by end user for specific reactive conditions.
Recommended Dilutions: ELISA: 1:450000; Western Blot: 1:5000 - 1:50000; Immunochemistry: 1:1000 - 1:5000;
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Mouse