Anti-Biotin Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
ROCK600-108-098
- Type antilichaam:Label
- Clonality:Polyklonaal
- Conjugatie:PE (fycoerythrin)
- ELISA:Yes
- Host:Goat
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- Maat:1 ml
- VPE:1 mL
Type: Primary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan