Accessories for wire baskets
Leverancier: BOCHEM
Ga naar:
Over dit artikel
6 Opties Hieronder Beschikbaar
Product Details & Documents
Product Family Options
Productinformatie
- BeschikbaarheidPrice
- BOCH17316275×375 mm, mesh dimensions: 10×10×3 mm
- BOCH17321275×375 mm, mesh dimensions: 20×20×3 mm
- BOCH17326275×375 mm, mesh dimensions: 35×35×3 mm
- BOCH17315575×375 mm, mesh dimensions: 10×10×3 mm
- BOCH17320575×375 mm, mesh dimensions: 20×20×3 mm
- BOCH17325575×375 mm, mesh dimensions: 35×35×3 mm