Bottles, coliform water sample
120 ml, Zonder thiosulfaat, PS, Case of 200
Catalogus # EPSC156-120ST
Leverancier: EP SCIENTIFIC
Bottles, coliform water sample
Catalogus # EPSC156-120ST
Leverancier: EP SCIENTIFIC
Ga naar:
Product Details & Documents
PP, PE or PS.
- Certified sterile; manufactured under an ISO certified quality management system
- Threaded screw closure for easy aseptic sample collection and secure cap engagement
- Non-autofluorescent
- Available pre-charged with a sodium thiosulfate (Na₂S₂O₃) tablet for sample de-chlorination, or without a tablet for un-chlorinated water sources
- 100 ml fill line
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.
Specificaties
- Verpakt:Case of 200
- Capaciteit:120 ml
- Materiaal:PS
- Seq:02
- Dosering:Zonder thiosulfaat
- Cat. No.:EPSC156-120ST
- Milieu certificering:NAR Certificate of Retirement
- Environmentally Preferable:Lage impact op de fabricage