Qualitative filter papers, slow filtration
The filter papers are used for very effective, fine filtration of small particles precipitates.
: Filters are supplied as flat in 100 filters boxes or folded from 100 mm Ø.
- Catalog No:
- PRATA015210
- PRATA007210
- PRATJ015210
- PRATA009210
- Gewicht:
- 100 g/m²
- 100 g/m²
- 100 g/m2
- 100 g/m²
- Dikte:
- 200 µm
- 200 µm
- 200 µm
- 200 µm
- Partikelretentie:
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
Frequently Bought Together