Orders LinkContactUsLinkComponent
Funnels, ribbed
Funnels, ribbed
80 mm, 18 mm, 100 mm
Catalog # KAVA632413004100
Supplier:   Kavalier
Funnels, ribbed
Catalog # KAVA632413004100
Supplier:  Kavalier
Funnels, ribbed
Funnels, ribbed
80 mm, 18 mm, 100 mm
Price
33,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With ribbed walls.
Specifications
  • Ø inw. boven:
    100 mm
  • Steel uitw. Ø:
    18 mm
  • Lengte steel:
    80 mm