Orders LinkContactUsLinkComponent
Latex tests
Latex tests
Test serum for Yersina enterocolitica identification, 1
Catalogus # LIOF96147
Leverancier:   LIOFILCHEM
Latex tests
Catalogus # LIOF96147
Leverancier:  LIOFILCHEM
CAS-nummer:  
Latex tests
Latex tests
Test serum for Yersina enterocolitica identification, 1
CAS-nummer:  
Price
260,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Identification of the main pathogenic microorganisms directly from clinical specimens.
  • Save time and labor with direct testing
  • Results in 18 to 24 hours
  • Confirmation with latex agglutination tests
  • Clear and well defined colour changes
  • Easy interpretation
Documents
Specificaties
Specificaties
  • VPE:
    1
  • Omschrijving:
    Test serum for Yersina enterocolitica identification
Specificaties