Racks for cryogenic vials, TruCool®
Catalog # BCISBCS-222
Supplier: BIOCISION
Specifications
- Aant. gaten:50 (5×10)
- Kleur:Veelkleurig
- Omschrijving:TruCool® cryogenic vial racks
- b×d×h:197×102×220 mm
- VPE:1 SET
Specifications
About this item
These polypropylene cryogenic racks feature a locking mechanism that allow one handed opening for self-standing cryogenic vials.
- Accommodates both round bottom and self-standing vial formats
- Racks have A to J row and 1 to 5 column indexing for easy organization
- Stackable when not in use
Multicolour pack contains one each of blue, green, orange, purple and pink racks.