Hypodermic needle
AIRTN224
- Meter:22
- Tip type:Bevelled
- Omschrijving:Hypodermic needle
- Lengte:102 mm
- VPE:100
Tri-bevel needles are constructed from the highest quality regular wall surgical stainless steel tubing available and meet rigorous quality control specifications. Ideal for veterinary and laboratory purpose.
: Packaged in individual sterile strip packs.