Anti-CD8A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K80]
USBIC2259-27U
- Type antilichaam:Primair
- antigeen symbool:CD8A
- Clonality:Monoklonaal
- Clone:5K80
- Conjugatie:PE (fycoerythrin)
- Flowcytometrie:Yes
- Host:Mouse
- ImmunoNeerslag:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Formulier:liquid
- Opslagbuffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Opslagtemperatuur:Cool
- Concentratie:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Maat:100 µg
- VPE:100 µG
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 5K80
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken