Order Entry
Belgium
ContactUsLinkComponent
Anti-Cardif Mouse Monoclonal Antibody [clone: Adri-1]
Anti-Cardif Mouse Monoclonal Antibody [clone: Adri-1]
Catalogus # USBI044625
Leverancier:  US Biological
Anti-Cardif Mouse Monoclonal Antibody [clone: Adri-1]
Catalogus # USBI044625
Leverancier:  US Biological

Specificaties

  • Type antilichaam:
    Primair
  • Clonality:
    Monoklonaal
  • Clone:
    Adri-1
  • Conjugatie:
    Onconjugeerd
  • Host:
    Mouse
  • ImmunoChemie:
    Yes
  • ImmunoNeerslag:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Formulier:
    liquid
  • Aminozuurgetal:
    160 to 450
  • Opslagbuffer:
    PBS, 10% glycerol, 0,02% sodium azide.
  • Opslagtemperatuur:
    Cool
  • Concentratie:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Maat:
    100 µg
  • VPE:
    100 µG

Specificaties

Over dit artikel

Type: Primary
Antigen: Cardif
Clonality: Monoclonal
Clone: Adri-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: