Anti-ZWILCH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI044427-FITC
- Type antilichaam:Primair
- antigeen symbool:ZWILCH
- Clonality:Polyklonaal
- Conjugatie:FITC (Fluoresceïne-isothiocyanaat)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitoop:C-terminal
- Formulier:liquid
- Aminozuurgetal:481 to 508
- Opslagbuffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Maat:200 µl
- VPE:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: ZWILCH (zwilch kinetochore protein)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse