Anti-MAPKAPK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBI038187-APC
- Type antilichaam:Primair
- antigeen symbool:MAPKAPK3
- Clonality:Polyklonaal
- Conjugatie:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitoop:N-terminal
- Formulier:liquid
- Aminozuurgetal:62 to 90
- Opslagbuffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Maat:200 µl
- VPE:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MAPKAPK3 (mitogen-activated protein kinase-activated protein kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse