Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 9G4.C3.F2]
Catalog # ROCK200-301-I42S
Supplier: Rockland Immunochemicals
Specifications
- Type antilichaam:Primair
- antigeen symbool:PROCALCITONIN
- Clonality:Monoklonaal
- Clone:9G4.C3.F2
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 lambda
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:none
- Formaat:Antibody fraction
- Formulier:Liquid (sterile filtered)
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Opslagtemperatuur:Cool
- Concentratie:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin Protein.
- Tested applications:Fluoresence ELISA
- Purification:Protein A purified
- Maat:25 µl
- VPE:25 µl
Specifications
About this item
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;
Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 9G4.C3.F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human