Anti-MORC3 Rabbit Polyclonal Antibody
Catalogus # ROCK100-401-N96S
Leverancier: Rockland Immunochemicals
Specificaties
- Type antilichaam:Primair
- antigeen symbool:MORC3
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:No
- Formaat:Antiserum
- Formulier:Liquid (sterile filtered)
- Gen-ID:338467
- Antigeen synoniemen:1110051N18Rik|D16Jhu32e|BF318192|Zcwcc3|AI452146|NXP2
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Opslagtemperatuur:Cool
- Concentratie:80 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogeen:Morc3 whole rabbit serum was prepared by repeated immunisations with a mouse Morc3 recombinant Protein
- Tested applications:Fluoresence ELISA
- Purification:delipidation and defibrination
- Maat:25 µl
- VPE:25 µl
Specificaties
Over dit artikel
Anti-Morc3 antibody is suitable for use in western blot. Specific conditions for reactivity should be optimised by the end user. Expect a band approximately 110.8 kDa in size corresponding Morc3 by western blotting in the appropriate cell lysate.
Recommended Dilutions: ELISA: 1:10000 to 1:50000; Western Blot: 1:1000 to 1:5000.
Type: Primary
Antigen: MORC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse