Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
ROCK807-4902
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:Texas Red®
- Host:Rabbit
- ImmunoFluorescentie:Yes
- Isotype:
- Reactivity:Golden syrian hamster
- Environmentally Preferable:
- Kruisadsorptie:none
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Opslagtemperatuur:Cool
- Concentratie:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Hamster IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Maat:500 µl
- VPE:500 µG
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Golden Syrian Hamster