Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # ROCK712-4026
Supplier: Rockland Immunochemicals
Specifications
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:TRITC (Tetramethylrhodamine-isothiocyanaat)
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Reactivity:Rat
- Western blot:Yes
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Antilichaamfragment:F(ab')2
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Opslagtemperatuur:Cool
- Concentratie:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Rat IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Maat:500 µl
- VPE:500 µG
Specifications
About this item
Secondary Rabbit Anti-IgG (H&L) Reacts with Rat
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat