Anti-IgG (H+L) Goat Polyclonal Antibody
ROCK711-1122
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Goat
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Isotype:
- Reactivity:Rabbit
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Liquid (sterile filtered)
- Antilichaamfragment:F(ab')2
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Opslagtemperatuur:Cool
- Concentratie:1,11 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Rabbit IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Maat:500 µg
- VPE:500 µG
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit