Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
Catalogus # ROCK710-4620
Leverancier: Rockland Immunochemicals
Specificaties
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:Biotine
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:Human
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Antilichaamfragment:F(ab')2
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Opslagtemperatuur:Cool
- Concentratie:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Mouse IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Maat:500 µg
- VPE:500 µG
Specificaties
Over dit artikel
This product has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse