Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Biotin)
Catalog # ROCK614-4604
Supplier: Rockland Immunochemicals
Specifications
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG F(ab')2
- Clonality:Polyklonaal
- Conjugatie:Biotine
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Isotype:
- Reactivity:Pig
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Opslagtemperatuur:Cool
- Concentratie:2,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Swine IgG F(ab')2 fragment
- Purification:purified by immunoaffinity chromatography
- Maat:2 mg
- VPE:2 mg
Specifications
About this item
Secondary Rabbit Anti-F(ab')2 Reacts with Pig (Swine, Porcine)
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Pig