Anti-IgM µ Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
ROCK610-4507
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgM µ
- Clonality:Polyklonaal
- Conjugatie:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoNeerslag:Yes
- Reactivity:Mouse
- Western blot:Yes
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Liquid (sterile filtered)
- Opslagbuffer:0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
- Opslagtemperatuur:Cool
- Concentratie:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Mouse IgM mu heavy chain
- Purification:purified by immunoaffinity chromatography
- Maat:1 mg
- VPE:1 mg
Anti-MOUSE IgM (alpha chain) (RABBIT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:20000;
Do not freeze!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Mouse