Anti-IgG (H+L) Chicken Polyclonal Antibody (Texas Red®)
Catalog # ROCK609-9002
Supplier: Rockland Immunochemicals
Specifications
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG (H+L)
- Clonality:Polyklonaal
- Conjugatie:Texas Red®
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Chicken
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Opslagtemperatuur:Cool
- Concentratie:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Human IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Maat:1 mg
- VPE:1 mg
Specifications
About this item
Secondary Chicken Anti-IgG (H&L) Reacts with Human
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgG
Reactivity: Human