Anti-IgA Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
ROCK609-1006
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgA
- Clonality:Polyklonaal
- Conjugatie:TRITC (Tetramethylrhodamine-isothiocyanaat)
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Goat
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Opslagbuffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Opslagtemperatuur:Cool
- Concentratie:2,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Human IgA alpha heavy chain
- Purification:purified by immunoaffinity chromatography
- Maat:1 mg
- VPE:1 mg
Secondary Goat Anti-IgA Reacts with Human
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Human