Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK602-4204
Supplier: Rockland Immunochemicals
Specifications
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG F(ab')2
- Clonality:Polyklonaal
- Conjugatie:FITC (Fluoresceïne-isothiocyanaat)
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Reactivity:Cat
- Western blot:Yes
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Formaat:Antibody fraction
- Formulier:Lyophilized
- Opslagbuffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Opslagtemperatuur:Cool
- Concentratie:2,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogeen:Cat IgG F(ab')2 fragment
- Purification:purified by immunoaffinity chromatography
- Maat:2 mg
- VPE:2 mg
Specifications
About this item
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Cat (Feline)
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Cat