Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalogus # ROCK308-4204
Leverancier: Rockland Immunochemicals
Specificaties
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG F(ab')2
- Clonality:Polyklonaal
- Conjugatie:FITC (Fluoresceïne-isothiocyanaat)
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Reactivity:Horse
- Western blot:Yes
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Maat:20 mg
- VPE:20 mg
Specificaties
Over dit artikel
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Horse