Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
50 mg
Catalogus # ROCK214-4104
Leverancier: Rockland Immunochemicals
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Catalogus # ROCK214-4104
Leverancier: Rockland Immunochemicals
Ga naar:
Product Details & Documents
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pig
Specificaties
- Maat:50 mg
- Western blot:Yes
- Clonality:Polyklonaal
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- antigeen symbool:IgG F(ab')2
- Type antilichaam:Ondergeschikt
- Host:Rabbit
- ChIP:Yes
- Shipping temperature:Ice
- Gelverschuiving:Yes
- ImmunoChemie:Yes
- Cat. No.:ROCK214-4104
- Conjugatie:Onconjugeerd
- Flowcytometrie:Yes
- Reactivity:Pig
- Neutralisation:Yes
- Opslagtemperatuur:Cool
- Kruisadsorptie:none
- ELISA:Yes



