Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK208-4204
Supplier: Rockland Immunochemicals
Specifications
- Type antilichaam:Ondergeschikt
- antigeen symbool:IgG F(ab')2
- Clonality:Polyklonaal
- Conjugatie:FITC (Fluoresceïne-isothiocyanaat)
- ELISA:Yes
- Flowcytometrie:Yes
- Gelverschuiving:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- ImmunoFluorescentie:Yes
- ImmunoNeerslag:Yes
- Reactivity:Horse
- Western blot:Yes
- Kruisadsorptie:none
- ChIP:Yes
- Neutralisation:Yes
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- Maat:20 mg
- VPE:20 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (Horse) (horse IgG)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Horse