Anti-EPEC Rabbit Polyclonal Antibody
Marketsource
Leverancier: Biorbyt
Over dit artikel
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: EPEC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
4 Opties Hieronder Beschikbaar
Product Details & Documents
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: EPEC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Specifications for Product Family
Specificaties
- Catalog No:
- BIRBORB13394-100
- BIRBORB13394-20
- BIRBORB13394-200
- BIRBORB13394-5
- antigeen symbool:
- EPEC
- EPEC
- EPEC
- EPEC
- Type antilichaam:
- Primair
- Primair
- Primair
- Primair
- Clonality:
- Polyklonaal
- Polyklonaal
- Polyklonaal
- Polyklonaal
- Conjugatie:
- Onconjugeerd
- Onconjugeerd
- Onconjugeerd
- Onconjugeerd
- Reactivity:
- Bacteria
- Bacteria
- Bacteria
- Bacteria
- Host:
- Rabbit
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- Yes
- Yes
- Immunogeen:
- Synthetic peptide
- Synthetic peptide
- Synthetic peptide
- Synthetic peptide
- Formulier:
- liquid
- liquid
- liquid
- liquid
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- purified by affinity chromatography
- purified by affinity chromatography
- Concentratie:
- 0,5 mg/ml
- 0,5 mg/ml
- Opslagbuffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Opslagtemperatuur:
- Cool
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice
- Ice
Specificaties
Product Family Options
Productinformatie
- MaatBeschikbaarheidPrice
- Specificaties:Marketsource
More
Specifications
Cat. No.BIRBORB13394-100antigeen symboolEPECType antilichaamPrimairClonalityPolyklonaalConjugatieOnconjugeerdReactivityBacteriaHostRabbitIsotypeIgGWestern blotYesImmunoChemieYesImmunogeenSynthetic peptideFormulierliquidPurificationpurified by affinity chromatographyConcentratie0,5 mg/mlOpslagbuffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolOpslagtemperatuurCoolShipping temperatureIceMarketsource - Specificaties:Marketsource
More
Specifications
Cat. No.BIRBORB13394-20antigeen symboolEPECType antilichaamPrimairClonalityPolyklonaalConjugatieOnconjugeerdReactivityBacteriaHostRabbitIsotypeIgGWestern blotYesImmunoChemieYesImmunogeenSynthetic peptideFormulierliquidPurificationpurified by affinity chromatographyConcentratieOpslagbuffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolOpslagtemperatuurCoolShipping temperatureIceMarketsource - Specificaties:Marketsource
More
Specifications
Cat. No.BIRBORB13394-200antigeen symboolEPECType antilichaamPrimairClonalityPolyklonaalConjugatieOnconjugeerdReactivityBacteriaHostRabbitIsotypeIgGWestern blotYesImmunoChemieYesImmunogeenSynthetic peptideFormulierliquidPurificationpurified by affinity chromatographyConcentratie0,5 mg/mlOpslagbuffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolOpslagtemperatuurCoolShipping temperatureIceMarketsource - Specificaties:Marketsource
More
Specifications
Cat. No.BIRBORB13394-5antigeen symboolEPECType antilichaamPrimairClonalityPolyklonaalConjugatieOnconjugeerdReactivityBacteriaHostRabbitIsotypeIgGWestern blotYesImmunoChemieYesImmunogeenSynthetic peptideFormulierliquidPurificationpurified by affinity chromatographyConcentratieOpslagbuffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolOpslagtemperatuurCoolShipping temperatureIceMarketsource



