Anti-ATP5J2 Rabbit Polyclonal Antibody
Catalog # ABGEAP5346C
Supplier: Abgent
Specifications
- Type antilichaam:Primair
- antigeen symbool:ATP5J2
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- ELISA:Yes
- Host:Rabbit
- ImmunoChemie:Yes
- Reactivity:Human
- Western blot:Yes
- Epitoop:middle
- Antigeen synoniemen:ATP5J2
- Opslagtemperatuur:Cool
- Shipping temperature:Ice
- VPE:400 µl
Specifications
About this item
Type: Primary
Antigen: ATP5J2 (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human