Pipette stands, Poxygrid®, SP Bel-Art
Catalog # BELAH189620001
Supplier: Bel-Art Products, a Part of SP
Specifications
- Voor:10 pipettes (Gilson Pipetman®); 2 pipettes with profile like Drummond Pipette-Aid®
- Kleur:blauw
- Omschrijving:For benchtop use
- Seq:01
- b×d×h:229×127×171 mm
- VPE:1
Specifications
About this item
These pipette stands made of epoxy coated aluminium provide storage and easy access for up to 10 Gilson Pipetman® pipettors plus up to two Pipet-Aid® pipettes.
- Keep pipettors organised, clearly visible and ready for use
- Epoxy coating is corrosion resistant and easy to clean
- Available with extended base with raised lip to hold plastic tip boxes
- For wall mouting or benchtop use