Qualitative filter papers, fast filtration
Supplier: PRAT DUMAS
Certificates
About this item
Filters on creped surface. Frequently used to determine the content of sucrose in cane sugar and in the pharmaceutical laboratories for common filtration. Also used for the industrial filtrations.
Ordering information: For specific orders, these filters can be supplied in other diameters or in sheets.
Packaging: Filters are supplied as flat in 100 filters boxes or folded from 150 mm of diameter.
Specifications
- Catalog No:
- PRATA009307
- PRATA015307
- PRATA007307
- PRATJ015307
- Gewicht:
- 75 g/m2
- 75 g/m2
- 75 g/m2
- 75 g/m2
- Dikte:
- 240 µm
- 240 µm
- 240 µm
- 240 µm
- Partikelretentie:
- 8 - 10 µm
- 8 - 10 µm
- 8 - 10 µm
- 8 - 10 µm
Specifications
Frequently Bought Together