Orders LinkContactUsLinkComponent
Digital stereo microscope, DM-143-FBGG-C
Supplier:   MOTIC
Digital stereo microscope, DM-143-FBGG-C
Supplier:  MOTIC
Certificates
Digital stereo microscope, DM-143-FBGG-C
Digital stereo microscope, DM-143-FBGG-C
4 Options Available Below
Product Details & Documents
This microscope features a binocular head, 45º inclined, with built-in digital camera (3 MP). It can be connected to a computer via USB port for viewing and capturing live images.
  • Greenough zoom optical system
  • Coarse focusing system with tension adjustment
  • Incident and transmitted halogen illumination with intensity control
  • Pole type stand with large working base
  • Large dual focusing knobs with slip clutch to prevent damage to mechanism
System requirements:
Windows XP or above, MAC OSX
Intel Pentium
1 Ghz processor, video card: 128 MB
RAM 512 MB or above
CD-ROM drive
USB 2.0
 : Supplied with software Motic Images Plus 2.0 for PC & MAC, dust cover and USB cable.
Product Family Options
Reticles
Product Information
  • Omschrijving
    Availability
    Price
  • Reticle with 140 divisions in 14 mm and crosshair (Ø = 23 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
  • Reticle with 70 divisions in 14 mm (Ø = 23 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
  • Reticle with 100 divisions in 10 mm (Ø = 23 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
  • Reticle with 360° protractor with 1° divisions and crosshair (Ø = 23 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
Eyepieces
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Widefield eyepiece, 15 mm
    15×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,10 €
    54,10 €
    1 * 1 ST
       54,10 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Widefield eyepiece, 10 mm
    20×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,60 €
    65,60 €
    1 * 1 ST
       65,60 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Micrometer eyepiece, 20 mm, 360° protractor with 1° divisions and crosshair
    10×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,80 €
    82,80 €
    1 * 1 ST
       82,80 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Micrometer eyepiece, 20 mm, 70 divisions in 14 mm
    10×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,80 €
    82,80 €
    1 * 1 ST
       82,80 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Widefield eyepiece, 20 mm
    10×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,60 €
    50,60 €
    1 * 1 ST
       50,60 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Micrometer eyepiece, 20 mm, 140 divisions in 14 mm and crosshair
    10×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,80 €
    82,80 €
    1 * 1 ST
       82,80 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Widefield eyepiece, 8 mm
    30×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,10 €
    85,10 €
    1 * 1 ST
       85,10 €
Product Information
  • Catalog #
    Omschrijving
    Vergroting
    Availability
    Price
  • Micrometer eyepiece, 20 mm, 100 divisions in 10 mm
    10×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,80 €
    82,80 €
    1 * 1 ST
       82,80 €
Accessories
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • 632-9038
    Polarisatieapparatuur B.301
    Log in for availability
    1 * 1 ST - 105,00 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • 632-9039
    Juwelenklem
    Log in for availability
    1 * 1 ST - 56,40 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • 632-9037
    Donkerveld-toevoeging met irisdiafragma
    Log in for availability
    1 * 1 ST - 113,00 €