Gas cylinder holder
Catalogus # 134-0000
Leverancier: R L ENTERPRISES
Specificaties
- Type:Cylinder holder, bench mounted
- VPE:1
Specificaties
Over dit artikel
This gas cylinder holder is made of heavy duty alloy casting that is secured to the benches up to 70 mm thick with two plated steel tommy screws. Two countersunk holes are provided to accept wood screws for permanent fixing.
- Suitable for gas cylinders up to 300 mm diameter
- Complete with fire resistant belt and buckle