Draining racks, wall mounted
Draining rack, with 24 rods, 20 arches with 2 base pole connectors, 420×170×610 mm
Catalog # KOTT359-00028
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Wire, coated.
- Good chemical resistance
- Good temperature resistance
- Easy to clean
Specifications
- Omschrijving:Draining rack, with 24 rods, 20 arches with 2 base pole connectors
- b×d×h:420×170×610 mm
- Cat. No.:KOTT359-00028