Pressure relief valves
VARIRE7030
: VARIAN
- Omschrijving:Valve, Rheodyne 7030, three-way switching, stainless steel, 1/16"
- VPE:1
Valves made from stainless steel are two-position with six-port. It is manually operated with three-way switching.