Power selector, WRT00117
Power selector, WRT00117
Catalogus # WINKWRT00117-XX010
Leverancier: WINKLER
Ga naar:
Product Details & Documents
Plug-in power selector made of polystyrene with switching mechanism.
- Rotary switch with bimetal element
- Signal light for controlling status indication
Specificaties
- Model:WRT00117
- Omschrijving:Power selector
- Cat. No.:WINKWRT00117-XX010
- b×d×h:120×57×56 mm
- Elektrisch:230 V; 10 A (ohmic load); 50 Hz