Orders LinkContactUsLinkComponent
Visual colour kits, glutaraldehyde
Glutaraldehyde test kit, model GT-1, colour disc, 0,5 - 2800 mg/l, 1 KIT
Catalogus # HACH2587200
Leverancier:   Hach
Visual colour kits, glutaraldehyde
Catalogus # HACH2587200
Leverancier:  Hach
CAS-nummer:  
Visual colour kits, glutaraldehyde
Visual colour kits, glutaraldehyde
Glutaraldehyde test kit, model GT-1, colour disc, 0,5 - 2800 mg/l, 1 KIT
CAS-nummer:  
Price
790,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Glutaraldehyde test kits are designed to monitor disinfection processes.
  • Simple, economical colour disc method
  • Accurate and quick results
  • Easy-to-carry test kit
 : Kits includes three reagents, colour discs, colour comparator box, bottles, sprinkler caps, syringes, cylinders, marker, ampoule breaker, ultra pure water, viewing tubes, instruction sheet, and carrying case.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    Glutaraldehyde test kit, model GT-1, colour disc
  • Meetbereik:
    0,5 - 2800 mg/l
Specificaties